SY3H3D67F516 Synthonix Corporation

2-Iodo-6-methoxyphenol

  • Cas Number : 111726-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45923.html

Safety Information

MSDS https://synthonix.com/45923.html
Availability details for
Quantity
  • Availability
  • Add to Cart