SY3H3D67F520 Synthonix Corporation

4-Iodo-N-methylaniline

  • MDL number No MDL found
  • Cas Number : 60577-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46120.html

Safety Information

MSDS https://synthonix.com/46120.html
Availability details for
Quantity
  • Availability
  • Add to Cart