SY3H3D67F52C Synthonix Corporation

Imidazo[1,2-a]pyridin-2(3H)-one hydrochloride

  • Cas Number : 52687-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46529.html

Safety Information

MSDS https://synthonix.com/46529.html
Availability details for
Quantity
  • Availability
  • Add to Cart