SY3H3D67F52E Synthonix Corporation

Indolin-4-amine

  • Cas Number : 52537-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46638.html

Safety Information

MSDS https://synthonix.com/46638.html
Availability details for
Quantity
  • Availability
  • Add to Cart