SY3H3D67F530 Synthonix Corporation

1-(4-Iodophenyl)ethanamine

  • MDL number No MDL found
  • Cas Number : 90086-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46768.html

Safety Information

MSDS https://synthonix.com/46768.html
Availability details for
Quantity
  • Availability
  • Add to Cart