SY3H3D67F532 Synthonix Corporation

4-Iodo-N,N-dimethylaniline

  • MDL number No MDL found
  • Cas Number : 698-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/46770.html

Safety Information

MSDS https://synthonix.com/46770.html
Availability details for
Quantity
  • Availability
  • Add to Cart