SY3H3D67F538 Synthonix Corporation

2-Iodo-4-(trifluoromethyl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 54507-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47477.html

Safety Information

MSDS https://synthonix.com/47477.html
Availability details for
Quantity
  • Availability
  • Add to Cart