SY3H3D67F53C Synthonix Corporation

6-Iodoquinazoline

  • MDL number No MDL found
  • Cas Number : 848841-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/47566.html

Safety Information

MSDS https://synthonix.com/47566.html
Availability details for
Quantity
  • Availability
  • Add to Cart