SY3H3D67F53F Synthonix Corporation

8-Iodoquinazolin-4(3H)-one

  • MDL number No MDL found
  • Cas Number : 77150-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47622.html

Safety Information

MSDS https://synthonix.com/47622.html
Availability details for
Quantity
  • Availability
  • Add to Cart