SY3H3D67F540 Synthonix Corporation

4-Iodo-2-(trifluoromethyl)benzamide

  • MDL number No MDL found
  • Cas Number : 1195677-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47756.html

Safety Information

MSDS https://synthonix.com/47756.html
Availability details for
Quantity
  • Availability
  • Add to Cart