SY3H3D67F542 Synthonix Corporation

4-Iodophthalic acid

  • Cas Number : 6301-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47827.html

Safety Information

MSDS https://synthonix.com/47827.html
Availability details for
Quantity
  • Availability
  • Add to Cart