SY3H3D67F543 Synthonix Corporation

Imidazo[1,5-a]pyridine-1-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 56671-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47871.html

Safety Information

MSDS https://synthonix.com/47871.html
Availability details for
Quantity
  • Availability
  • Add to Cart