SY3H3D67F544 Synthonix Corporation

4-(Imidazo[1,2-a]pyrazin-2-yl)-1,2,5-oxadiazol-3-amine

  • Cas Number : 1031927-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47941.html

Safety Information

MSDS https://synthonix.com/47941.html
Availability details for
Quantity
  • Availability
  • Add to Cart