SY3H3D67F546 Synthonix Corporation

4-Iodo-1H-indole

  • MDL number No MDL found
  • Cas Number : 81038-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47984.html

Safety Information

MSDS https://synthonix.com/47984.html
Availability details for
Quantity
  • Availability
  • Add to Cart