SY3H3D67F548 Synthonix Corporation

5-Iodo-2-methylbenzonitrile

  • MDL number No MDL found
  • Cas Number : 52107-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47986.html

Safety Information

MSDS https://synthonix.com/47986.html
Availability details for
Quantity
  • Availability
  • Add to Cart