SY3H3D67F54D Synthonix Corporation

6-Iodo-1-methyl-1H-indazole

  • Cas Number : 1214899-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48347.html

Safety Information

MSDS https://synthonix.com/48347.html
Availability details for
Quantity
  • Availability
  • Add to Cart