SY3H3D67F54F Synthonix Corporation

3-Iodo-7-methoxy-1H-indazole

  • MDL number No MDL found
  • Cas Number : 351210-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48381.html

Safety Information

MSDS https://synthonix.com/48381.html
Availability details for
Quantity
  • Availability
  • Add to Cart