SY3H3D67F55B Synthonix Corporation

2-Iodo-4-methoxybenzoic acid

  • MDL number No MDL found
  • Cas Number : 54435-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48689.html

Safety Information

MSDS https://synthonix.com/48689.html
Availability details for
Quantity
  • Availability
  • Add to Cart