SY3H3D67F562 Synthonix Corporation

1H-Indole-6-sulfonamide

  • MDL number No MDL found
  • Cas Number : 145951-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48831.html

Safety Information

MSDS https://synthonix.com/48831.html
Availability details for
Quantity
  • Availability
  • Add to Cart