SY3H3D67F564 Synthonix Corporation

1-Iodo-2,3-dimethylbenzene

  • MDL number No MDL found
  • Cas Number : 31599-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49168.html

Safety Information

MSDS https://synthonix.com/49168.html
Availability details for
Quantity
  • Availability
  • Add to Cart