SY3H3D67F56B Synthonix Corporation

4-Iodo-1,2-dimethoxybenzene

  • MDL number No MDL found
  • Cas Number : 5460-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49399.html

Safety Information

MSDS https://synthonix.com/49399.html
Availability details for
Quantity
  • Availability
  • Add to Cart