SY3H3D67F56E Synthonix Corporation

4-Iodo-2,6-dimethylphenol

  • MDL number No MDL found
  • Cas Number : 10570-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49470.html

Safety Information

MSDS https://synthonix.com/49470.html
Availability details for
Quantity
  • Availability
  • Add to Cart