SY3H3D67F573 Synthonix Corporation

Imidazo[1,2-a]pyridine-5-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 1198283-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49596.html

Safety Information

MSDS https://synthonix.com/49596.html
Availability details for
Quantity
  • Availability
  • Add to Cart