SY3H3D67F575 Synthonix Corporation

(1H-Indazol-5-yl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 943845-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49623.html

Safety Information

MSDS https://synthonix.com/49623.html
Availability details for
Quantity
  • Availability
  • Add to Cart