SY3H3D67F580 Synthonix Corporation

3-Iodo-6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazole

  • Cas Number : 1309788-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50316.html

Safety Information

MSDS https://synthonix.com/50316.html
Availability details for
Quantity
  • Availability
  • Add to Cart