SY3H3D67F581 Synthonix Corporation

1H-Indole-2-carbonitrile

  • MDL number No MDL found
  • Cas Number : 36193-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50792.html

Safety Information

MSDS https://synthonix.com/50792.html
Availability details for
Quantity
  • Availability
  • Add to Cart