SY3H3D67F582 Synthonix Corporation

4-Iodoisoquinoline

  • Cas Number : 55270-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50860.html

Safety Information

MSDS https://synthonix.com/50860.html
Availability details for
Quantity
  • Availability
  • Add to Cart