SY3H3D67F584 Synthonix Corporation

6-Iodoquinolin-2-ol

  • MDL number No MDL found
  • Cas Number : 99455-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50959.html

Safety Information

MSDS https://synthonix.com/50959.html
Availability details for
Quantity
  • Availability
  • Add to Cart