SY3H3D67F586 Synthonix Corporation

4-Iodoisoquinolin-1-amine

  • MDL number No MDL found
  • Cas Number : 55270-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/51097.html

Safety Information

MSDS https://synthonix.com/51097.html
Availability details for
Quantity
  • Availability
  • Add to Cart