SY3H3D67F58B Synthonix Corporation

Indolizine-6-carboxylic acid

  • Cas Number : 588720-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51178.html

Safety Information

MSDS https://synthonix.com/51178.html
Availability details for
Quantity
  • Availability
  • Add to Cart