SY3H3D67F58D Synthonix Corporation

3-(3-Iodophenyl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 41070-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51272.html

Safety Information

MSDS https://synthonix.com/51272.html
Availability details for
Quantity
  • Availability
  • Add to Cart