SY3H3D67F58F Synthonix Corporation

2-(1H-Imidazol-2-yl)phenol

  • MDL number No MDL found
  • Cas Number : 52755-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51353.html

Safety Information

MSDS https://synthonix.com/51353.html
Availability details for
Quantity
  • Availability
  • Add to Cart