SY3H3D67F591 Synthonix Corporation

1-(Imidazo[1,2-a]pyridin-3-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 29096-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51361.html

Safety Information

MSDS https://synthonix.com/51361.html
Availability details for
Quantity
  • Availability
  • Add to Cart