SY3H3D67F597 Synthonix Corporation

3-Iodo-6-methoxy-1-methyl-1H-indazole

  • MDL number No MDL found
  • Cas Number : 1431163-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/51700.html

Safety Information

MSDS https://synthonix.com/51700.html
Availability details for
Quantity
  • Availability
  • Add to Cart