SY3H3D67F598 Synthonix Corporation

7-Iodo-6-methoxy-4-methyl-1H-indazole

  • MDL number No MDL found
  • Cas Number : 1352397-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51701.html

Safety Information

MSDS https://synthonix.com/51701.html
Availability details for
Quantity
  • Availability
  • Add to Cart