SY3H3D67F59B Synthonix Corporation

1H-Indole-5-carbohydrazide

  • MDL number No MDL found
  • Cas Number : 406192-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51713.html

Safety Information

MSDS https://synthonix.com/51713.html
Availability details for
Quantity
  • Availability
  • Add to Cart