SY3H3D67F5A0 Synthonix Corporation

2-Iodo-3,4-dimethylbenzoic acid

  • MDL number No MDL found
  • Cas Number : 129833-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51965.html

Safety Information

MSDS https://synthonix.com/51965.html
Availability details for
Quantity
  • Availability
  • Add to Cart