SY3H3D67F5A4 Synthonix Corporation

2-(Imidazo[1,2-a]pyridin-3-yl)acetic acid hydrochloride

  • Cas Number : 127657-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52221.html

Safety Information

MSDS https://synthonix.com/52221.html
Availability details for
Quantity
  • Availability
  • Add to Cart