SY3H3D67F5A9 Synthonix Corporation

(4-Imidazol-1-yl-phenyl)methanol

  • MDL number No MDL found
  • Cas Number : 86718-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52295.html

Safety Information

MSDS https://synthonix.com/52295.html
Availability details for
Quantity
  • Availability
  • Add to Cart