SY3H3D67F5AD Synthonix Corporation

Inosine 5'-monophosphate disodium salt hydrate

  • MDL number No MDL found
  • Cas Number : 352195-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52535.html

Safety Information

MSDS https://synthonix.com/52535.html
Availability details for
Quantity
  • Availability
  • Add to Cart