SY3H3D67F5B3 Synthonix Corporation

1-Iodo-4-isopropyl-2-methoxybenzene

  • MDL number No MDL found
  • Cas Number : 1240304-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/53021.html

Safety Information

MSDS https://synthonix.com/53021.html
Availability details for
Quantity
  • Availability
  • Add to Cart