SY3H3D67F5B8 Synthonix Corporation

1H-Indole-2,5-dicarboxylic acid

  • Cas Number : 117140-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53583.html

Safety Information

MSDS https://synthonix.com/53583.html
Availability details for
Quantity
  • Availability
  • Add to Cart