SY3H3D67F5BB Synthonix Corporation

3-(1-Imidazolyl)benzaldehyde

  • Cas Number : 127404-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53641.html

Safety Information

MSDS https://synthonix.com/53641.html
Availability details for
Quantity
  • Availability
  • Add to Cart