SY3H3D67F5BE Synthonix Corporation

4-(1H-Imidazol-2-yl)benzoic acid

  • MDL number MFCD00456057
  • Cas Number : 108035-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53658.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53658.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart