SY3H3D67F5C8 Synthonix Corporation

1-(4-Iodophenyl)piperidin-2-one

  • MDL number No MDL found
  • Cas Number : 385425-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/54536.html

Safety Information

MSDS https://synthonix.com/54536.html
Availability details for
Quantity
  • Availability
  • Add to Cart