SY3H3D67F5CA Synthonix Corporation

(1H-Indol-3-yl)-1-propanamine

  • MDL number No MDL found
  • Cas Number : 6245-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54694.html

Safety Information

MSDS https://synthonix.com/54694.html
Availability details for
Quantity
  • Availability
  • Add to Cart