SY3H3D67F5CC Synthonix Corporation

(Z)-3-(1H-Indol-3-yl)acrylonitrile

  • MDL number No MDL found
  • Cas Number : 85452-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55220.html

Safety Information

MSDS https://synthonix.com/55220.html
Availability details for
Quantity
  • Availability
  • Add to Cart