SY3H3D67F5CD Synthonix Corporation

2-(1H-Indol-3-yl)-2-methylpropanoic acid

  • Cas Number : 2770-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55818.html

Safety Information

MSDS https://synthonix.com/55818.html
Availability details for
Quantity
  • Availability
  • Add to Cart