SY3H3D67F5D0 Synthonix Corporation

(S)-1-(4-Iodo-5-methoxy-2-nitrophenyl)-2,2-dimethylpropan-1-ol

  • Cas Number : 1427311-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56062.html

Safety Information

MSDS https://synthonix.com/56062.html
Availability details for
Quantity
  • Availability
  • Add to Cart