SY3H3D67F5D1 Synthonix Corporation

4-((4-Iodobenzyl)oxy)piperidine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1220034-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56070.html

Safety Information

MSDS https://synthonix.com/56070.html
Availability details for
Quantity
  • Availability
  • Add to Cart